Methylprednisolone acetate

Proveedor: TCI

M2635-1G M2635-250MG
TCIAM2635-1GEA 201 EUR
TCIAM2635-1G TCIAM2635-250MG
Methylprednisolone acetate
Methylprednisolone acetate
Formula: C₂₄H₃₂O₆
Número MDL: MFCD00200356
Núm. CAS: 53-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR