Methanesulphonylacetic acid

Proveedor: TCI

Sinónimos: 2-(Methylsulphonyl)acetic acid

M2770-1G M2770-5G
TCIAM2770-1GEA 38 EUR
TCIAM2770-1G TCIAM2770-5G
Methanesulphonylacetic acid
Methanesulphonylacetic acid
Formula: C₃H₆O₄S
MW: 138.14 g/mol
Número MDL: MFCD00051843
Núm. CAS: 2516-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR