2-Methoxy-5-sulphamoylbenzoic acid

Proveedor: TCI

M2771-25G M2771-5G
TCIAM2771-25GEA 132 EUR
TCIAM2771-25G TCIAM2771-5G
2-Methoxy-5-sulphamoylbenzoic acid
2-Methoxy-5-sulphamoylbenzoic acid
Formula: C₈H₉NO₅S
Número MDL: MFCD00129997
Núm. CAS: 22117-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR