Mangiferin

Proveedor: TCI

M2886-1G M2886-5G
TCIAM2886-1GEA 60 EUR
TCIAM2886-1G TCIAM2886-5G
Mangiferin
Mangiferin
Formula: C₁₉H₁₈O₁₁
Número MDL: MFCD00075656
Núm. CAS: 4773-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR