Methyl 5-formyl-2-methoxybenzoate

Proveedor: TCI

M3327-25G M3327-5G
TCIAM3327-25GEA 165 EUR
TCIAM3327-25G TCIAM3327-5G
Methyl 5-formyl-2-methoxybenzoate
Methyl 5-formyl-2-methoxybenzoate
Formula: C₁₀H₁₀O₄
MW: 194,184 g/mol
Número MDL: MFCD10565654
Núm. CAS: 78515-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR