4-Methoxybutanoic acid

Proveedor: TCI

M3329-1G M3329-250MG
TCIAM3329-1GEA 144 EUR
TCIAM3329-1G TCIAM3329-250MG
4-Methoxybutanoic acid
4-Methoxybutanoic acid
Formula: C₅H₁₀O₃
Número MDL: MFCD00029855
Núm. CAS: 29006-02-8
UN: 3265

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR