2-Metoxi-5-nitrobenzaldehído

Proveedor: TCI

N1034-25G N1034-5G
TCIAN1034-25GEA 127 EUR
TCIAN1034-25G TCIAN1034-5G
2-Metoxi-5-nitrobenzaldehído
2-Metoxi-5-nitrobenzaldehído
Formula: C₈H₇NO₄
Número MDL: MFCD02093683
Núm. CAS: 25016-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR