NS-1619
Proveedor: TCI
Sinónimos:
1-(2-Hydroxy-5-(trifluoromethyl)phenyl)-5-(trifluoromethyl)-1,3-dihydro-2H-benzo[d]imidazol-2-one
N1191-10MG
N1191-50MG
TCIAN1191-10MGEA
119
EUR
TCIAN1191-10MG
TCIAN1191-50MG
NS-1619
NS-1619
Formula:
C₁₅H₈F₆N₂O₂ MW: 362,2 g/mol |
Número MDL:
MFCD00928190 Núm. CAS: 153587-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...