Oxybutynin hydrochloride

Proveedor: TCI

O0451-1G O0451-5G
TCIAO0451-1GEA 67 EUR
TCIAO0451-1G TCIAO0451-5G
Oxybutynin hydrochloride
Oxybutynin hydrochloride
Formula: C₂₂H₃₁NO₃·HCl
Número MDL: MFCD00072150
Núm. CAS: 1508-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR