Sodium (S)-6-methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulphinyl)benzo[d]imidazol-1-ide
Proveedor: TCI
Sinónimos:
Esomeprazole sodium, (S)-Omeprazole sodium salt
O0560-25G
O0560-5G
TCIAO0560-25GEA
232
EUR
TCIAO0560-25G
TCIAO0560-5G
Sodium (S)-6-methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulphinyl)benzo[d]imidazol-1-ide
Sodium (S)-6-methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulphinyl)benzo[d]imidazol-1-ide
Formula:
C₁₇H₁₈N₃NaO₃S |
Número MDL:
MFCD07802816 Núm. CAS: 161796-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...