3-Oxo-3-(2-thienyl)propionitrile

Proveedor: TCI

O0571-25G O0571-5G
TCIAO0571-25GEA 272 EUR
TCIAO0571-25G TCIAO0571-5G
3-Oxo-3-(2-thienyl)propionitrile
3-Oxo-3-(2-thienyl)propionitrile
Formula: C₇H₅NOS
Número MDL: MFCD00052586
Núm. CAS: 33898-90-7
UN: 3439

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR