Palmatine chloride

Proveedor: TCI

Sinónimos: 2,3,9,10-Tetramethoxy-5,6-dihydroisoquinolino[3,2-a]isoquinolin-7-ium chloride

P2138-1G P2138-5G
TCIAP2138-1GEA 116 EUR
TCIAP2138-1G TCIAP2138-5G
Palmatine chloride
Palmatine chloride
Formula: C₂₁H₂₂ClNO₄
MW: 387,86 g/mol
Número MDL: MFCD00016656
Núm. CAS: 10605-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR