Pseudopelletierine

Proveedor: TCI

Sinónimos: 9-Methyl-9-azabicyclo[3.3.1]nonan-3-one

P2648-1G P2648-5G
TCIAP2648-1GEA 68 EUR
TCIAP2648-1G TCIAP2648-5G
Pseudopelletierine
Pseudopelletierine
Formula: C₉H₁₅NO
MW: 153,23 g/mol
Número MDL: MFCD00078161
Núm. CAS: 552-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR