Potassium tetraphenylborate

Proveedor: TCI

P2661-1G P2661-5G
TCIAP2661-1GEA 61 EUR
TCIAP2661-1G TCIAP2661-5G
Potassium tetraphenylborate
Potassium tetraphenylborate
Formula: C₂₄H₂₀BK
Número MDL: MFCD03093877
Núm. CAS: 3244-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR