2-(5H-Chromeno[2,3-b]pyridin-7-yl)propanoic acid

Proveedor: TCI

P2778-1G P2778-5G
TCIAP2778-1GEA 120 EUR
TCIAP2778-1G TCIAP2778-5G
2-(5H-Chromeno[2,3-b]pyridin-7-yl)propanoic acid
2-(5H-Chromeno[2,3-b]pyridin-7-yl)propanoic acid
Formula: C₁₅H₁₃NO₃
Número MDL: MFCD00866111
Núm. CAS: 52549-17-4
UN: 2811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR