6-Quinolineboronic acid (Contains varying amounts of Anhydride)
Proveedor: TCI
Sinónimos:
Quinolin-6-ylboronic acid, Quinoline-6-boronic acid
Q0098-1G
Q0098-5G
TCIAQ0098-1GEA
55
EUR
TCIAQ0098-1G
TCIAQ0098-5G
6-Quinolineboronic acid (Contains varying amounts of Anhydride)
6-Quinolineboronic acid
Formula:
C₉H₈BNO₂ |
Número MDL:
MFCD03093100 Núm. CAS: 376581-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...