Sodium 3,5,6-trichloropyridin-2-olate

Proveedor: TCI

T3004-1G T3004-5G
TCIAT3004-1GEA 32 EUR
TCIAT3004-1G TCIAT3004-5G
Sodium 3,5,6-trichloropyridin-2-olate
Sodium 3,5,6-trichloropyridin-2-olate
Formula: C₅HCl₃NNaO
MW: 220.41 g/mol
Número MDL: MFCD08704561
Núm. CAS: 37439-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR