N-Methyl-3-(triethoxysilyl)propylamine
Proveedor: TCI
Sinónimos:
N-Methyl-3-(triethoxysilyl)propan-1-amine, [3-(Methylamino)prop-1-yl]triethoxysilane
T3242-25G
T3242-5G
TCIAT3242-25GEA
863
EUR
TCIAT3242-25G
TCIAT3242-5G
N-Methyl-3-(triethoxysilyl)propylamine
N-Methyl-3-(triethoxysilyl)propylamine
Formula:
C₁₀H₂₅NO₃Si MW: 235,396 g/mol |
Número MDL:
MFCD24842713 Núm. CAS: 6044-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...