4-(p-Tolyloxy)benzoic acid

Proveedor: TCI

T3391-1G T3391-5G
TCIAT3391-1GEA 69 EUR
TCIAT3391-1G TCIAT3391-5G
4-(p-Tolyloxy)benzoic acid
4-(p-Tolyloxy)benzoic acid
Formula: C₁₄H₁₂O₃
Número MDL: MFCD01631893
Núm. CAS: 21120-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR