Triamcinolone diacetate

Proveedor: TCI

T3676-250MG T3676-50MG
TCIAT3676-250MGEA 192 EUR
TCIAT3676-250MG TCIAT3676-50MG
Triamcinolone diacetate
Triamcinolone diacetate
Formula: C₂₅H₃₁FO₈
Número MDL: MFCD00056462
Núm. CAS: 67-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR