4-[(Trimethylsilyl)ethynyl]benzaldehyde

Proveedor: TCI

T3827-1G T3827-5G
TCIAT3827-1GEA 44 EUR
TCIAT3827-1G TCIAT3827-5G
4-[(Trimethylsilyl)ethynyl]benzaldehyde
4-[(Trimethylsilyl)ethynyl]benzaldehyde
Formula: C₁₂H₁₄OSi
Número MDL: MFCD02093765
Núm. CAS: 77123-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR