4-(2H-Tetrazol-5-yl)benzoic acid

Proveedor: TCI

T3917-1G T3917-200MG
TCIAT3917-1GEA 303 EUR
TCIAT3917-1G TCIAT3917-200MG
4-(2H-Tetrazol-5-yl)benzoic acid
4-(2H-Tetrazol-5-yl)benzoic acid
Formula: C₈H₆N₄O₂
Número MDL: MFCD01569290
Núm. CAS: 34114-12-0

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR