2-(3-(Thiophene-2-carbonyl)phenyl)propanoic acid
Proveedor: TCI
Sinónimos:
Suprofen, 2-(4-(Thiophene-2-carbonyl)phenyl)propanoic acid
U0138-100MG
U0138-25MG
TCIAU0138-100MGEA
355
EUR
TCIAU0138-100MG
TCIAU0138-25MG
2-(3-(Thiophene-2-carbonyl)phenyl)propanoic acid
2-(3-(Thiophene-2-carbonyl)phenyl)propanoic acid
Formula:
C₁₄H₁₂O₃S MW: 260.30 g/mol |
Número MDL:
MFCD00079572 Núm. CAS: 40828-46-4 UN: 2811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...