CleanPrint 10 Cleanroom Papers
Proveedor: VELTEK ASSOCIATES
CLP10-A4-02 CLP10-8.5X1102 CLP108.5113H02 CLP10-A4-01 CLP108.5BL3H02 CLP108.5X11BL1 CLP108.5X11BL2 CLP108.5X11GR1 CLP108.5X11GR2 CLP108.5X11YL1 CLP108.5X11YL2
VELTCLP10-A4-02EA 2620 EUR
VELTCLP10-A4-02 VELTCLP10-8.5X1102 VELTCLP108.5113H02 VELTCLP10-A4-01 VELTCLP108.5BL3H02 VELTCLP108.5X11BL1 VELTCLP108.5X11BL2 VELTCLP108.5X11GR1 VELTCLP108.5X11GR2 VELTCLP108.5X11YL1 VELTCLP108.5X11YL2
CleanPrint 10 Cleanroom Papers
Papeles
CleanPrint 10 papers are low particulate and non-shedding synthetic writing substrates for use in cleanrooms.
  • Lightweight and flexible but exceptionally durable
  • Excellent ability to write
  • Inks adhere and dry immediately
  • Abrasion, chemical, water, tear and smear resistant
  • Completely traceable from start to finish
  • Sterile papers are gamma irradiated to 10⁻⁶ SAL
These papers remain pliable in extreme temperatures of –70 to +180 °C. These are cellulose free and extremely low in particulates and shedding features. Reams are individually labelled with lot number and expiration.
Certificaciones: Sterile versions are lot sterility tested according to current USP compendium.
Envase: Packed as 100 sheets per ream, 5 reams per case with 500 total sheets. Sterile papers are packed in double bag.