Lacosamide

Proveedor: Thermo Fisher Scientific

465321000 465325000
465321000.EA 129 EUR
465321000. 465325000.
Lacosamide
Lacosamide

Order Now

Resultados test especificaciones

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Water ≤0.5 %
Specific optical rotation +14° to +18° (20°C, 589 nm) (0.1g in 10 ml CH3OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR