2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate 96%
Proveedor: Thermo Fisher Scientific
|
Atención
|
470730010
470730050
470730010.EA
61.7
EUR
470730010.
470730050.
2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate 96%
2-(Methacryloyloxy)ethyl-2-(trimethylammonio)ethyl phosphate
Formula:
C₁₁H₂₂NO₆P Temperatura de almacenaje: Nevera |
Número MDL:
MFCD11112180 Núm. CAS: 67881-98-5 |
Resultados test especificaciones
Appearance | white to light yellow crystalline powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥95 % |
Titration with HClO4 | 95 to 105 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...