Drospirenone 98%
Proveedor: Thermo Fisher Scientific
|
Peligro
|
Sinónimos:
Dihydrospirorenone
472040010
472042500
472040010.EA
201
EUR
472040010.
472042500.
Drospirenone 98%
Drospirenone
Formula:
C₂₄H₃₀O₃ MW: 366.50 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00867350 Núm. CAS: 67392-87-4 EINECS: 266-679-2 UN: 3077 ADR: 9 III Índice Merck: 13,03488 |
Resultados test especificaciones
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 199°C to 203°C |
Specific optical rotation | -187° to -193° (20°C, 589 nm) (0.1g in 10 ml methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...