Shellac gum

Proveedor: Thermo Fisher Scientific

473460010 473462500
473460010.EA 269 EUR
473460010. 473462500.
Shellac gum
Shellac gum
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00148309
Núm. CAS: 9000-59-3
EINECS: 232-549-9
Índice Merck: 13,08553

Order Now

Resultados test especificaciones

Appearance yellow to brown flakes
Infrared spectrum Conforms
Loss on drying ≤2.0 % (105°C, 3 hrs)
Solubility (20% in Ethanol) Clear to light turbid solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR