RosetteSep™ Human CD45 Depletion Cocktails
Proveedor: STEMCELL Technologies
15162 15122
STMC15162EA 1193 EUR
STMC15162 STMC15122
RosetteSep™ Human CD45 Depletion Cocktails
Medios para separación celular
Immunodensity depletion of CD45+ cells.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human CD45 Depletion Cocktail is designed to enrich epithelial circulating tumor cells (CTCs) from whole blood by depleting CD45+ cells. Unwanted cells are targeted for depletion with Tetrameric Antibody Complexes recognising CD45, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The purified epithelial tumor cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Precaución: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).