EasySep™ Human Pan-ILC Enrichment Kits
Proveedor: STEMCELL Technologies
17975 17975RF
STMC17975EA 1182 EUR
STMC17975 STMC17975RF
EasySep™ Human Pan-ILC Enrichment Kits
Medios para separación celular
Immunomagnetic enrichment of untouched human ILC1, ILC2, and ILC3s.
  • Fast, easy-to-use and column-free
  • Untouched, viable cells
  • Facilitates rapid flow sorting of ILCs
The EasySep™ Human Pan-ILC Enrichment Kit is designed to enrich Group 1, 2, and 3 Innate Lymphoid Cells (ILC1, 2, and 3) from leukapheresis products by immunomagnetic negative selection. Unwanted cells are targeted for removal by Tetrameric Antibody Complexes directed against non-ILCs and dextran-coated magnetic particles (RapidSpheres™). labelled cells are separated using an EasySep™ magnet without the use of columns. Unwanted cells remain in the tube as enriched ILCs are simply poured off into a new tube. Isolated cells are immediately available for flow sorting and other downstream applications.
Precaución: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').