Verde luz SF, powder or crystals, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY

Sinónimos: Dihidrogeno(etil)[4-[p-[etil(m-sulfonatobencil)amino]-p'-sulfonatobencidrilen]ciclohexa-2,5-dien-1-iliden](m-sulfonatobencil)amonio sal de disodio

M-Clarity™ quality level = MQ100

L5382-100G L5382-10G L5382-25G
L5382-100GEA 811 EUR
L5382-100G L5382-10G L5382-25G
Verde luz SF, powder or crystals, Sigma-Aldrich®
Verde luz SF

Light green SF yellowish (LGSFY) is a disodium salt of an acid triphenylmethane dye. It is also known as Light Green, Lissamine Green SF, Food Green 2, Green No. 205, Acid Green, Acid Brilliant Green 5, and Acid Brilliant Green SF. LGSFY is an important component of Papanicolaou stains in addition to eosin Y and bismark brown Y. Light green SF yellowish is a histological stain for collagen and a permitted colorant for foodstuffs and beverages.


Light green SF is generally used as a cytoplasmic counterstain with nuclear stains like safranine and Feulgen stain. In histopatology, it is used for collagen fiber staining in Masson′s trichrome, Papanicolaou cytological polychrome stains, and Twort stain for microorganisms in tissue sections. It is a constituent of polychrome staining method used for differentiating secretory cells of the pituitary gland. Light green SF also stains the cellulose cell walls of plant cells.

Formula: C₃₇H₃₄N₂Na₂O₉S₃
Pto. de fusión: 288 °C (desc., lit.)
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00012121
Núm. CAS: 5141-20-8
EINECS: 225-906-5
Índice Merck: 13,05506
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ba2cb5842fa7e7b291da25832612e8d008429d2&vid=87ee50ef45f4d5807095d6e17e48be90962866a2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance (Colour) Dark Red to Purple
Appearance (Form) Powder or Crystals
Nitrogen 2.1 - 3.5 %
Sodium (Na) ≥5%
Solubility (Colour) Dark Blue-Green to Very Dark Blue-Green to Very Dark Green and Dark Brown
Solubility (Turbidity), 10 mg/ml, H₂O Clear
E1% ≥600
Wavelength in H₂O 631 - 634 nm

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR