Bidones, Nalgene®
Proveedor: Thermo Fisher Scientific
2240-0015 2240-0025 2240-0050
216-8691EA 82.8 EUR
216-8691 216-8692 216-8693
Bidones, Nalgene®
Frascos Bidones
HDPE, translúcidos con tapón de rosca rosca de PP blanco.
  • Buena resistencia química
  • Resistente a temperaturas desde −100 hasta +120 °C
  • Diseño estable y robusto, con graduación en galones y litros; herméticos
Good stability through a large base and low centre of gravity. The spout is long enough for accurate pouring of liquids. With tethered sealing cap that cannot be lost. Recessed base offers additional support during pouring.