Safety waste funnels, Nalgene®
Proveedor: Thermo Fisher Scientific
6378-0004 6378-0010
119-3034EA 83.5 EUR
119-3034 119-3040
Safety waste funnels, Nalgene®
Embudos Embudos de seguridad
HDPE funnels and covers, with PP closure and funnel adapter, PTFE vent membrane.
  • Vent system helps reduce fumes and splashing
  • Removable neck screen traps large particles
  • Available in 10" Ø funnel with 83B closure and 5¹/₂" funnel with 38 to 430 closure
For solvent, chemical and biological waste. Features the easy snap safety latch to minimise spillage if accidentally tipped over and a larger diameter funnel for easy pouring.