Bottle top vacuum filtration systems, cellulose nitrate membrane, Rapid-Flow™ Nalgene®
Proveedor: Thermo Fisher Scientific
121-0020 121-0045 125-0020 125-0045 125-0080 126-0020 126-0045 126-0080 127-0020 127-0045 127-0080 450-0045 450-0080 450-0020
734-5001EA 140 EUR
734-5001 734-5002 734-5003 734-5004 734-5005 734-5006 734-5007 734-5008 734-5009 734-5010 734-5011 734-5083 734-5084 734-5082
Bottle top vacuum filtration systems, cellulose nitrate membrane, Rapid-Flow™ Nalgene®
Filtros Filtros de la parte superior de botellas
Rapid-Flow™ disposable filter units with cellulose nitrate membrane, PS funnel and collection reservoir feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput.
  • Color coded according to membrane
  • Membrane with excellent wetting properties and fastest flow rates with aqueous solutions
  • Funnel and receiver flask graduated
  • Quick disconnect tubing adapter with cover
Sterile disposable filters for cold sterilisation, clarification, prefiltration and ultrafiltration of buffers, culture media and aqueous solutions.
More Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Certificación ambiental ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement
IRECS; ISO 14001 (Certificate Registration No. EMS 752603) NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)