Discos de filtro
Proveedor: KARTELL
835 836 837 838 839 840 843 844
519-5001EA 11.4 EUR
519-5001 519-5002 519-5003 519-5004 519-5005 519-5006 519-5007 519-5008
Discos de filtro
Filtros Discos de filtros
HDPE filter discs for use between filter paper and perforated Büchner plate to prevent clogging of filter holes.