Forceps, straight
Proveedor: USBECK Laborgeräte
3020 3021 3022 3023 3024 3025 3035 3033 3034 3030 3031 3032
232-0094EA 3.4 EUR
232-0094 232-0095 232-0096 232-0097 232-0098 232-0099 USBE3035 USBE3033 USBE3034 USBE3030 USBE3031 USBE3032
Forceps, straight
Pinzas
Made from nickel plated steel or from stainless steel.
  • Straight
  • Fine points