Pipetas graduadas, BLAUBRAND®, Tipo 2, clase AS, vaciado total
Proveedor: Brand
27816 27817 27818 27819 27820 27821 27822 27823 27824 27825 27826 27827 27828 27829
612-3151EA 12 EUR
612-3151 612-3152 612-3153 612-3154 612-3156 612-3157 612-3158 612-3159 612-3161 612-3162 612-3663 612-3664 612-3666 612-4758
Pipetas graduadas, BLAUBRAND®, Tipo 2, clase AS, vaciado total
Pipetas Pipetas graduadas
AR-Glas®.
  • Calibrated to deliver (TD, Ex), type 2: Nominal volume on top
  • Marks and inscriptions in high contrast blue enamel
  • Waiting time 5 seconds
  • Includes a batch certificate
  • Also available (on request) with an individual certificate, USP individual certificate or DAkks calibration certificate
DIN EN ISO 835.
Longitud: 360 mm ±10 mm, excepto las pipetas de 25/50 ml: 450 mm ±10 mm
Certificaciones: DIN EN ISO 835. Certificado de conformidad.
Precaución: * With cotton plug end
** In addition to the ISO range
*** InMore Product Information