Pipetas graduadas, con pistón
Proveedor: HIRSCHMANN
1190133 1190141 1190153 1190160 1190165 1190170
612-1025EA 24.55 EUR
612-1025 612-1026 612-1027 612-1028 612-1029 612-1030
Pipetas graduadas, con pistón
Pipetas Pipetas graduadas
AR-Glas®.
  • Calibrated to deliver ‘Ex’
  • Durable amber stain graduation, main point ring graduation
  • Integrated syringe pipetting aid