Puntas con filtro
Proveedor: Brand
732712 732714 732722 732724 732726 732728 732730 732732 732734 732706 732610 732602 732604 732710 732608 732708 732612 732614 732622 732624 732626 732628 732630 732632 732634 732702 732704 732606
613-2378EA 58.7 EUR
613-2378 613-2379 613-2380 613-2381 613-2382 613-2383 613-2384 613-2385 613-2386 613-2375 613-2363 613-2358 613-2359 613-2377 613-2362 613-2376 613-2364 613-2365 613-2366 613-2367 613-2368 613-2369 613-2370 613-2371 613-2372 613-2373 613-2374 613-2361
Puntas con filtro
Puntas de pipetas
These filter tips have a PE filter that is free from chemical additives and therefore prevents contamination of the samples. Permeability is controlled by the combination of pore size and filter length, so that no aerosols can reach the pipette shaft. The filters function with consistent reliability. However, if liquids accidently contact the filter, they pass very slowly. Since the filter does not swell, the sample can be recovered from the filter by simply actuating the pipette's blow-out function, or by centrifugation if necessary.
  • High-purity polypropylene, free from DiHEMDA and oleamide, manufactured without lubricants; cadmium-free pigments
  • Rack-packed pipette tips are free from DNA, RNases, endotoxins and ATP, independent of the packaging
  • Graduations for quick volume checking
  • Sterile tips and packaging are manufactured exclusively under BIO-CERT® quality certification
  • Autoclavable at +121 °C (2 bar), acc. DIN EN 285
The tips are tested for compatibility with BRAND pipettes and are suitable for use with most of the common pipettor brands including Gilson®, Thermo Scientific Finnpipette®, Eppendorf® and Sartorius.
Bulk packaging in reclosable bags, non sterile
Tips are produced under supervised cleanroom conditions and automatically shrink wrapped in reclosable bags and packaged in cardboard boxes. The batch number is printed on every bag.
TipBox, non sterile and sterile, BIO-CERT®
Tip box with transparent window, hinged and push-on lid (dual function closure) and coloured tip trays, with side panel labelling. Usable with a partially loaded multi channel pipette.
TipRack, non sterile and sterile, BIO-CERT®
Space saving refill unit for TipBox, with an environmentally compatible packaging made of recyclable PET. Compared to the filled TipBoxes, the amount of waste is reduced by 20%. Sterile racks are supplied with a transfer aid that enables simple, contamination-free transfer into a previously sterilised TipBox. All tip-trays are printed on one side with information on the contents.
BIO-CERT®: These tips are made from high-quality, pure PP without any mold release agents and other additives. All lots are tested for sterility (acc. ISO 11 137 and AAMI guidelines, a SAL of 10⁻⁶ is obtained) and are certified free from endotoxins, DNA, RNase and ATP. Tips are supplied with a certificate of analysis.