Draining racks, Azlon®
Proveedor: SciLabware
WRP001P WRP002P
212-7548EA 398 EUR
212-7548 AZLOWRP002P
Draining racks, Azlon®
Gradillas Soportes para drenaje
Easy to assemble, lightweight polystyrene draining racks designed to be wall mounted.
  • Pegs slot into place on the back board and are spaced to allow various sizes of lab ware to fit on to the rack
  • Water is collected in the trough at the base of the rack and drains away via a rubber tube which can be directed into a nearby sink or container
Ø/peg: 16 mm
Información de suministro: Supplied with wall mounting accessories.