Distilling bridges, Claisen
Proveedor: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
05055201 05055301 05055304 05055404
271-3141EA 101 EUR
271-3141 271-3142 201-1133 271-3144
Distilling bridges, Claisen
Componentes para destilación Puentes
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Still heads with Liebig condenser and Claisen attachment. Socket for distillation thermometer with 55 mm total length. Liebig condenser with straight outlet. Connection for coolant water via tube connector.