Sterican® Single-Use Needles for Special Applications
Proveedor: B. Braun
4665643 4665635 4665120B 4665791B 4665317B
612-0161EA 38.7 EUR
612-0161 612-0162 BRAU4665120B BRAU4665791B BRAU4665317B
Sterican® Single-Use Needles for Special Applications
Agujas de jeringas
Stainless, chromium-nickel steel with transparent PP Luer lock plastic hub. These sterile hypodermic needles Sterican® have the following features: Suitable for single-use hypodermic needles for intra-muscular, subcutaneous, intravenous, intra-arterial and special injections. Thin walled needles.
  • Extremely smooth surface with light silicone coating
  • 3-facet tips, long or short bevel
  • For minimal pain upon puncture
Certificaciones: ISO 6009