VWR®, Dry Block/Incubator Thermometers
Proveedor: VWR Collection
620-0814EA 111 EUR
620-0814 620-0815
VWR®, Dry Block/Incubator Thermometers
Termómetros Analogue Thermometers
Designed for use in all standard heating and drying blocks, water baths and incubators. Contains blue organic liquid fill against lead-free white back glass.
  • El recubrimiento opcional de PTFE evita la contaminación del líquido y el vidrio
  • Inmersión parcial
  • Números de serie individuales
Immersion depth: 35 mm
Certificaciones: Traceable to NIST.
Información de suministro: Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy indicating accuracy traceable to NIST.