Glass tubing
Proveedor: EMMANUEL PIERRE
1TAR910 1TAR1010 1TAR410 1TAR510 1TAR610 1TAR710 1TAR810
212-0205EA 26.2 EUR
212-0205 212-0199 212-0200 212-0201 212-0202 212-0203 212-0204
Glass tubing
Tuberías Tubo de transferencia
AR-glass.
  • Good chemical and physical properties
  • Suitable for the production of items that must normally withstand only brief exposure to aggressive chemicals
  • Not resistant to high thermal loads
Length: 500 mm.