Estereomicroscopios, ST-30/ST-36
Proveedor: MOTIC
51020141 PS30425201 PS36425201 PS3045B201 PS3645B201
632-9018EA 50.6 EUR
632-9018 630-3015 630-0691 MOTIPS3045B201 MOTIPS3645B201
Estereomicroscopios, ST-30/ST-36
Microscopios
These stereo microscopes are designed for education as well as quality control.
  • Bright, sharp image without heat
  • Magnification is changed by turning the turret
  • Comfortable long-term use, low consumption, environmental choice
  • Rear-facing head, forward-facing head or 360° rotatable head
  • Efficient LED energy saving lamp
Información de suministro: Supplied with frosted glass stage plates and dust cover.
Order Now
ESPECIFICACIONES
Modelo ST-30C-2LOO ST-36C-2LOO ST-30C-6LED ST-36C-6LED
Environmentally Preferable
Cabezal 45° de inclinación 45° de inclinación 45° de inclinación 45° de inclinación
Oculares Gran campo 10×/20 mm Gran campo 10×/20 mm
Objetivos 2×, 4× 2×, 4× 2×, 4× 2×, 4×
Iluminación Halógena Halógena LED LED