Deioniser with changeable cartridge
Proveedor: AQUADEM
REGE800S DEM-E100-001 DEM-E200-001 DEM-E300-001 DEM-E500-001 DEM-E600-001 E080 E200 E300 E500 E600 PMEFP210 PMEFP310 REGE100S REGE200S REGE300S REGE500S REGE600S E100
AQUDREGE800SEA 969 EUR
AQUDREGE800S 171-0100 171-0101 171-0102 171-0103 171-0104 171-0106 171-0107 171-0108 171-0109 171-0110 171-0122 171-0123 AQUDREGE100S AQUDREGE200S AQUDREGE300S AQUDREGE500S AQUDREGE600S 171-0337
Deioniser with changeable cartridge
Purificación de agua Desionizadores
The polyamide range is a set of demineralisation columns using mixed-bed ion exchange resin technology. These devices ensure deionised water production of between 5 and 15 MΏ/cm according to whether the installation type is single or double cycle.
  • L×D×H: 173×102×663 mm
  • Connections: ¾ inch.
Connections: ¾ inch.
These filtration panels are accessories for demineralisation cylinders specifically designed for use in laboratories or health facilities. The cylinders ensure optimal retention of the mineral salts dissolved in the feed water, in order to obtain purified water of a very high quality.
Información accesorios: Filtration panels
These filtration panels are accessories for demineralisation cylinders specifically designed for use in laboratories or health facilities. The cylinders ensure optimal retention of the mineral salts dissolved in the feed water, in order to obtain purified water of a very high quality.
With 2 or 3 filters.
One filtration panel comprises a flow control valve, a pressure regulator, an upstream pressure gauge, a 5 µm CA prefilter, a digital resistivity meter, a 1 µm final filter, a downstream pressure gauge and 2 bleed valves.