Indigocarmín, dark powder

Proveedor: MP Biomedicals
Atención

Sinónimos: CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue

0215263805 0215263880
ICNA0215263805EA 30.5 EUR
ICNA0215263805 ICNA0215263880
Indigocarmín, dark powder
Indigocarmin

Indigo Carmine is a blue, anionic indigoid dye.
Indigo Carmine is useful in the staining of Negri bodies. It can be used as a contrast stain for plasma when red nuclear stains are used.


  • Presentation: Dark Powder
  • Soluble in water (3 mg/mL) and ethanol (3 mg/mL).
  • Storage temperature: Store at room Temperature, protect from light.

Formula: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00005723
Núm. CAS: 860-22-0
EINECS: 212-728-8
Índice Merck: 13,04969

Order Now

Resultados test especificaciones

Identity Test Passes
UV/Vis: l max (water) 605-615nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR