Lantano, composto con níquel (1:5) ≥99.9% (REO, rare earth oxide basis), REacton®

Proveedor: Thermo Scientific

Sinónimos: Lanthanum nickel

11271.18 11271.90
11271.18EA 134 EUR
11271.18 11271.90
Lantano, composto con níquel (1:5) ≥99.9% (REO, rare earth oxide basis), REacton®
Lantano, composto con níquel (1:5)

Lanthanum nickel act as a superconducting intermetallic phase in lanthanum nickel boro-nitride.


Purity: 99.9% CAS No.: 12196-72-4 Molecular Formula: LaNi5 Physical Form: Pieces, Intermetallic, Packaged under argon Sensitivity: Air and Moisture Sensitive Odour: Odourless UN No.: 3178 MDL No.: MFCD00198177

Formula: LaNi₅
MW: 432,36 g/mol
Número MDL: MFCD00198177
Núm. CAS: 12196-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR