Intramedic™ medical/surgical tubing
Proveedor: DIFCO BD
427416 427415 427406 427400 427420 427425 427430 427435 427440 427445 427401 427411 427421 427426 427431 427436 427441 427446 427451 427519 427450
228-1867EA 0 EUR
228-1867 228-1859 228-1869 228-1861 228-1858 228-1857 228-1856 228-1855 228-1854 228-1851 228-1870 228-1868 228-1852 228-1866 228-1865 228-1850 228-1864 228-1863 228-1862 228-1253 228-1853
Intramedic™ medical/surgical tubing
Tuberías Tubo de transferencia
PE, translucent.
  • Durable over a wide temperature range, up to 104°C
  • Waxy white, translucent, tasteless, odorless
  • Polyethylene contains no additives or plasticizers
This tubing is non-radiopaque and has non-tissue-reacting properties. Rigid, but flexible enough for applications using vacuum and pressure.