Ácido palmítico ≥99%, en polvo

Proveedor: MP Biomedicals

Sinónimos: Ácido hexadecanoico

0210090583 0210090510 0210090580
ICNA0210090583EA 733 EUR
ICNA0210090583 ICNA0210090510 ICNA0210090580
Ácido palmítico ≥99%, en polvo
Ácido palmítico

Palmitic acid, or hexadecanoic acid is one of the most common 16 carbon saturated fatty acids found in animals and plants. It occurs as the glyceryl ester in many oils and fats


It has been reported to target proteins to cell membranes. It has been found to promote triglyceride accumulation and also affect cell viability. Triglyceride accumulation in goose hepatocytes shows the ability to induce apoptosis.


Mechanistic studies show that Palmitic Acid has also demonstrated the ability to induce opening of Ca2+-dependent pores in plasma membranes of red blood cells which will eventually lead to lysis of the cells. Additional studies suggest that saturated fatty acids can induce the expression of Cox-2 and NFκB (nuclear factor κB) activation. Palmitic acid is used in determination of water hardness.


Soluble in DMF (0,5% w/v - clear and colourless); cold alcohol (sparingly), petroleum ether (sparingly), hot alcohol (freely), ether (freely), propyl alcohol (freely), and chloroform (freely). Insoluble in water.

Formula: C₁₆H₃₂O₂
MW: 256,43 g/mol
Pto. Ebullición: 352 °C (1013 hPa)
Pto. de fusión: 61…63 °C
Densidad: 0,853 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00002747
Núm. CAS: 57-10-3
EINECS: 200-312-9
Índice Merck: 13,07064

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR